missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIAL1 Protein
A cDNA sequence encoding the TIAL1 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | TIAL1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934579
|
enQuireBio™
QP13749-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924579
|
enQuireBio™
QP13749-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914579
|
enQuireBio™
QP13749-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TIAL1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TIAL1 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol, 2mM DTT and 1mM EDTA . |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMEDDGQ PRTLYVGNLS RDVTEVLILQ LFSQIGPCKS CKMITEHTSN DPYCFVEFYE HRDAAAALAA MNGRKILGKE VKVNWATTPS SQKKDTSNHF HVFVGDLSPE ITTEDIKSAF APFGKISDAR VVKDMATGKS KGYGFVSFYN KLDAENAIVH MGGQWLGGRQ IRTNWATRKP PAPKSTQENN TKQLRFEDVV NQSSPKNCTV YCGGIASGLT DQLMRQTFSP FGQIMEIRVF PEKGYSFVRF STHESAAHAI VSVNGTTIEG HVVKCYWGKE SPDMTKNFQQ VDYSQWGQWS QVYGNPQQYG QYMANGWQVP PYGVYGQPWN QQGFGVDQSP SAAWMGGFGA QPPQGQAPPP VIPPPNQAGY GMASYQTQ | |
Greater than 90.0% as determined by SDS-PAGE. |