missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TIMM17A Protein

A DNA sequence encoding the Homo sapiens (Human) TIMM17A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6791-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005890

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006326.1
TIMM17A Protein
Research Use Only
TIMM17A
Recombinant Protein
E. coli
MEEYAREPCP WRIVDDCGGA FTMGTIGGGI FQAIKGFRNS PVGVNHRLRG SLTAIKTRAP QLGGSFAVWG GLFSMIDCSM VQVRGKEDPW NSITSGALTG AILAARNGPV AMVGSAAMGG ILLALIEGAG ILLTRFASAQ FPNGPQFAED PSQLPSTQLP SSPFGDYRQY Q
Greater than 80% as determined by SDS-PAGE.
10440
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers