missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM17A Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM17A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6791-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006326.1 | |
TIMM17A Protein | |
Research Use Only | |
TIMM17A | |
Recombinant Protein | |
E. coli | |
MEEYAREPCP WRIVDDCGGA FTMGTIGGGI FQAIKGFRNS PVGVNHRLRG SLTAIKTRAP QLGGSFAVWG GLFSMIDCSM VQVRGKEDPW NSITSGALTG AILAARNGPV AMVGSAAMGG ILLALIEGAG ILLTRFASAQ FPNGPQFAED PSQLPSTQLP SSPFGDYRQY Q | |
Greater than 80% as determined by SDS-PAGE. |
10440 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |