missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Titin Protein

A DNA sequence encoding the Homo sapiens (Human) Titin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8823-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16026671

  • 7550.00 SEK / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001243779.1
Titin Protein
Research Use Only
TTN
Recombinant Protein
E. coli
VGSSICSARV TLREPPSFIK KIESTSSLRG GTAAFQATLK GSLPITVTWL KDSDEITEDD NIRMTFENNV ASLYLSGIEV KHDGKYVCQA KNDAGIQRCS ALLSVKEPAT ITEEAVSIDV TQGDPATLQV KFSGTKEITA KWFKDGQELT LGSKYKISVT DTVSILKIIS TEKKDSGEYT FEVQNDVGRS SCKARINVLD LIIPPSF
Greater than 90% as determined by SDS-PAGE.
7273
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers