missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFR1 / CD120a / TNFRSF1A Protein
A DNA sequence encoding the Homo sapiens (Human) TNFR1 / CD120a / TNFRSF1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8566-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001056.1 | |
TNFR1 / CD120a / TNFRSF1A Protein | |
Research Use Only | |
TNFRSF1A | |
Recombinant Protein | |
E. coli | |
VPHLGDREKR DSVCPQGKYI HPQNNSICCT KCHKGTYLYN DCPGPGQDTD CRECESGSFT ASENHLRHCL SCSKCRKEMG QVEISSCTVD RDTVCGCRKN QYRHYWSENL FQCFNCSLCL NGTVHLSCQE KQNTVCTCHA GFFLRENECV SCSNCKKSLE CTKLCLPQIE NVKGTEDSGT | |
Greater than 90% as determined by SDS-PAGE. |
7132 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |