missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF11A Protein
A DNA sequence encoding the Homo sapiens (Human) TNFRSF11A, was expressed in the hosts and tags indicated.
2405.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_001257878.1 |
---|---|
Gene ID (Entrez) | 8792 |
Name | TNFRSF11A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16065521
|
enQuireBio™
QP8604-EC-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055521
|
enQuireBio™
QP8604-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005531
|
enQuireBio™
QP8604-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085521
|
enQuireBio™
QP8604-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095521
|
enQuireBio™
QP8604-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075521
|
enQuireBio™
QP8604-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257878.1 | |
TNFRSF11A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
8792 | |
Research Use Only | |
TNFRSF11A | |
Recombinant Protein | |
E. coli | |
LQIAPPCTSE KHYEHLGRCC NKCEPGKYMS SKCTTTSDSV CLPCGPDEYL DSWNEEDKCL LHKVCDTGKA LVAVVAGNST TPRRCACTAG YHWSQDCECC RRNTECAPGL GAQHPLQLNK DTVCKPCLAG YFSDAFSSTD KCRPWTNCTF LGKRVEHHGT EKSDAVCSSS LPARK | |
Greater than 90% as determined by SDS-PAGE. |