missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF10 / TRAIL / APO-2L Protein
A DNA sequence encoding the Homo sapiens (Human) TNFSF10 / TRAIL / APO-2L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6817-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001177871.1 | |
TNFSF10 / TRAIL / APO-2L Protein | |
Research Use Only | |
TNFSF10 | |
Recombinant Protein | |
E. coli | |
TNELKQMQDK YSKSGIACFL KEDDSYWDPN DEESMNSPCW QVKWQLRQLV RKMILRTSEE TISTVQEKQQ NISPLVRERG PQRVAAHITG TRGRSNTLSS PNSKNEKALG RKINSWESSR SGHSFLSNLH LRNGELVIHE KGFYYIYSQT YFRFQEEIKE NTKNDKQMVQ YIYKYTSYPD PILLMKSARN SCWSKDAEYG LYSIYQGGIF ELKENDRIFV SVTNEHLIDM DHEASFFGAF LVG | |
Greater than 90% as determined by SDS-PAGE. |
8743 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |