Learn More
enQuireBio™ Recombinant Human TNFSF12 Protein
A DNA sequence encoding the Homo sapiens (Human) TNFSF12, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6819-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16076060
  • 26990.00 SEK / 1mg
Estimated Shipment: 16-07-2024
to see stock.
Specifications
NP_003800.1
TNFSF12 Protein
Research Use Only
TNFSF12
Recombinant Protein
E. coli
SLGSRASLSA QEPAQEELVA EEDQDPSELN PQTEESQDPA PFLNRLVRPR RSAPKGRKTR ARRAIAAHYE VHPRPGQDGA QAGVDGTVSG WEEARINSSS PLRYNRQ
Greater than 90% as determined by SDS-PAGE.
407977
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol