missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFSF12 Protein

A DNA sequence encoding the Homo sapiens (Human) TNFSF12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6819-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056060

  • 5980.00 SEK / 100µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_003800.1
TNFSF12 Protein
Research Use Only
TNFSF12
Recombinant Protein
E. coli
SLGSRASLSA QEPAQEELVA EEDQDPSELN PQTEESQDPA PFLNRLVRPR RSAPKGRKTR ARRAIAAHYE VHPRPGQDGA QAGVDGTVSG WEEARINSSS PLRYNRQ
Greater than 90% as determined by SDS-PAGE.
407977
100 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers