missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Toll-like receptor 8 Protein
A DNA sequence encoding the Homo sapiens (Human) Toll-like receptor 8, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9437-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057694.2 | |
Toll-like receptor 8 Protein | |
Research Use Only | |
TLR8 | |
Recombinant Protein | |
Yeast | |
EENFSRSYPC DEKKQNDSVI AECSNRRLQE VPQTVGKYVT ELDLSDNFIT HITNESFQGL QNLTKINLNH NPNVQHQNGN PGIQSNGLNI TDGAFLNLKN LRELLLEDNQ LPQIPSGLPE SLTELSLIQN NIYNITKEGI SRLINLKNLY LAWNCYFNKV CEKTNIEDGV FETLTNLELL SLSFNSLSHV PPKLPSSLRK LFLSNTQIKY ISEEDFKGLI NLTLLDLSGN CPRCFNAPFP CVPCDGGASI NIDRFAFQNL TQLRYLNLSS TSLRKINAAW FKNMPHLKVL DLEFNYLVGE IASGAFLTML PRLEILDLSF NYIKGSYPQH INISRNFSKL LSLRALHLRG YVFQELREDD FQPLMQLPNL STINLGINFI KQIDFKLFQN FSNLEIIYLS ENRISPLVKD TRQSYANSSS FQRHIRKRRS TDFEFDPHSN FYHFTRPLIK PQCAAYGKAL DLSLNSIFFI GPNQFENLPD IACLNLSANS NAQVLSGTEF SAIPHVKYLD LTNNRLDFDN ASALTELSDL EVLDLSYNSH YFRIAGVTHH LEFIQNFTNL KVLNLSHNNI YTLTDKYNLE SKSLVELVFS GNRLDILWND DDNRYISIFK GLKNLTRLDL SLNRLKHIPN EAFLNLPASL TELHINDNML KFFNWTLLQQ FPRLELLDLR GNKLLFLTDS LSDFTSSLRT LLLSHNRISH LPSGFLSEVS SLKHLDLSSN LLKTINKSAL ETKTTTKLSM LELHGNPFEC TCDIGDFRRW MDEHLNVKIP RLVDVICASP GDQRGKSIVS LELTTCVSDV T | |
Greater than 90% as determined by SDS-PAGE. |
51311 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |