missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription elongation factor A protein 1
A DNA sequence encoding the Homo sapiens (Human) Transcription elongation factor A protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7582-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003186.1 | |
Transcription elongation factor A protein 1 | |
Research Use Only | |
TCEA2 | |
Recombinant Protein | |
E. coli | |
MMGKEEEIAR IARRLDKMVT KKSAEGAMDL LRELKAMPIT LHLLQSTRVG MSVNALRKQS SDEEVIALAK SLIKSWKKLL DASDAKARER GRGMPLPTSS RDASEAPDPS RKRPELPRAP STPRITTFPP VPVTCDAVRN KCREMLTAAL QTDHDHVAIG ADCERLSAQI EECIFRDVGN TDMKYKNRVR SRISNLKDAK NPDLRRNVLC GAITPQQIAV MTSEEMASDE LKEIRKAMTK EAIREHQMAR TGGTQTDLFT CGKCRKKNCT YTQVQTRSSD EPMTTFVVCN ECGNRWKFC | |
Greater than 90% as determined by SDS-PAGE. |
6919 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |