missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Trichoplein keratin filament-binding Protein
A DNA sequence encoding the Homo sapiens (Human) Trichoplein keratin filament-binding protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8130-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137324.1 | |
Trichoplein keratin filament-binding Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
84260 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TCHP | |
Recombinant Protein | |
E. coli | |
MALPTLPSYW CSQQRLNQQL ARQREQEARL RQQWEQNSRY FRMSDICSSK QAEWSSKTSY QRSMHAYQRE KMKEEKRRSL EARREKLRQL MQEEQDLLAR ELEELRLSMN LQERRIREQH GKLKSAKEEQ RKLIAEQLLY EHWKKNNPKL REMELDLHQK HVVNSWEMQK EEKKQQEATA EQENKRYENE YERARREALE RMKAEEERRQ LEDKLQAEAL LQQMEELKLK EVEATKLKKE QENLLKQRWE LERLEEERKQ MEAFRQKAEL GRFLRHQYNA QLSRRTQQIQ EELEADRRIL QALLEKEDES QRLHLARREQ VMADVAWMKQ AIEEQLQLER AREAELQMLL REEAKEMWEK REAEWARERS ARDRLMSEVL TGRQQQIQEK IEQNRRAQEE SLKHREQLIR NLEEVRELAR REKEESEKLK SARKQELEAQ VAERRLQAWE ADQQEEEEEE EARRVEQLSD ALLQQEAETM AEQGYRPKPY GHPKIAWN |