missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Trimethyllysine dioxygenase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Trimethyllysine dioxygenase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6808-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055990

  • 26990.00 SEK / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001171726.1
Trimethyllysine dioxygenase, mitochondrial Protein
Research Use Only
TMLHE
Recombinant Protein
E. coli
LLKGGVIYPA LPQPNFKSLL PLAVHWHHTA SKSLTCAWQQ HEDHFELKYA NTVMRFDYVW LRDHCRSASC YNSKTHQRSL DTASVDLCIK PKTIRLDETT LFFTWPDGHV TKYDLNWLVK NSYEGQKQKV IQPRILWNAE IYQQAQVPSV DCQSFLETNE GLKKFLQNFL LYGIAFVENV PPTQEHTEKL AERISLIRET IYGRMWYFTS DFSRGDTAYT KLALDRHTDT TYFQEPCGIQ VFHCLKHEGT GGRTLLVDGF YAAEQVLQKA PEEFELLSKV PLKHEYIEDV GECHNHMIGI GPVLNIYPWN KELYLIRLFK EKQNTVNRQW NSSLQCDIPE RILTYRHFVS GTSIEHRGSL I
Greater than 90% as determined by SDS-PAGE.
55217
1 mg
Not determined.
Full Length of Isoform 4
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers