missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TSLP Protein

A DNA sequence encoding the Homo sapiens (Human) TSLP, was expressed in the hosts and tags indicated.

6835.00 SEK - 19130.00 SEK

Specifications

Accession Number NP_149024.1
Gene ID (Entrez) 85480
Name TSLP Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16026250
View Documents Promotion Details
enQuireBio™
QP6845-MA-20UG
20 μg
6835.00 SEK
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 22-01-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16016250
View Documents Promotion Details
enQuireBio™
QP6845-MA-100UG
100 μg
19130.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 22-01-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16036250
View Documents Promotion Details
enQuireBio™
QP6845-MA-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_149024.1
TSLP Protein
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
85480
Research Use Only
TSLP
Recombinant Protein
Mammal
YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications