missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tubulin beta-2A chain Protein

A DNA sequence encoding the Homo sapiens (Human) Tubulin beta-2A chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6852-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056290

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001060.1
Tubulin beta-2A chain Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
7280
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TUBB2A
Recombinant Protein
Yeast
MREIVHIQAG QCGNQIGAKF WEVISDEHGI DPTGSYHGDS DLQLERINVY YNEAAGNKYV PRAILVDLEP GTMDSVRSGP FGQIFRPDNF VFGQSGAGNN WAKGHYTEGA ELVDSVLDVV RKESESCDCL QGFQLTHSLG GGTGSGMGTL LISKIREEYP DRIMNTFSVM PSPKVSDTVV EPYNATLSVH QLVENTDETY SIDNEALYDI CFRTLKLTTP TYGDLNHLVS ATMSGVTTCL RFPGQLNADL RKLAVNMVPF PRLHFFMPGF APLTSRGSQQ YRALTVPELT QQMFDSKNMM AACDPRHGRY LTVAAIFRGR MSMKEVDEQM LNVQNKNSSY FVEWIPNNVK TAVCDIPPRG LKMSATFIGN STAIQELFKR ISEQFTAMFR RKAFLHWYTG EGMDEMEFTE AESNMNDLVS EYQQYQDATA DEQGEFEEEE GEDEA
Videos
SDS
Documents