missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin polymerization-promoting Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin polymerization-promoting protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6830-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_008961.1 | |
Tubulin polymerization-promoting Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
11076 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
TPPP | |
Recombinant Protein | |
E. coli | |
MADKAKPAKA ANRTPPKSPG DPSKDRAAKR LSLESEGAGE GAAASPELSA LEEAFRRFAV HGDARATGRE MHGKNWSKLC KDCQVIDGRN VTVTDVDIVF SKIKGKSCRT ITFEQFQEAL EELAKKRFKD KSSEEAVREV HRLIEGKAPI ISGVTKAISS PTVSRLTDTT KFTGSHKERF DPSGKGKGKA GRVDLVDESG YVSGYKHAGT YDQKVQGGK |