missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tumor protein D52 Protein
A DNA sequence encoding the Homo sapiens (Human) Tumor protein D52, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6825-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020423.1 | |
Tumor protein D52 Protein | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
7163 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TPD52 | |
Recombinant Protein | |
E. coli | |
MDRGEQGLLR TDPVPEEGED VAATISATET LSEEEQEELR RELAKVEEEI QTLSQVLAAK EKHLAEIKRK LGINSLQELK QNIAKGWQDV TATSAYKKTS ETLSQAGQKA SAAFSSVGSV ITKKLEDVKN SPTFKSFEEK VENLKSKVGG TKPAGGDFGE VLNSAANASA TTTEPLPEKT QESL |