missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tumor suppressor candidate 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Tumor suppressor candidate 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6856-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056330

  • 6685.00 SEK / 200µg


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_009206.1
Tumor suppressor candidate 2 Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
11334
Greater than 80% as determined by SDS-PAGE.
Research Use Only
TUSC2
Recombinant Protein
E. coli
GASGSKARGL WPFASAAGGG GSEAAGAEQA LVRPRGRAVP PFVFTRRGSM FYDEDGDLAH EFYEETIVTK NGQKRAKLRR VHKNLIPQGI VKLDHPRIHV DFPVILYEV
Videos
SDS
Documents