missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Twinfilin-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Twinfilin-2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_009215.1 |
---|---|
Gene ID (Entrez) | 11344 |
Name | Twinfilin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080721
|
enQuireBio™
QP7777-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16000731
|
enQuireBio™
QP7777-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090721
|
enQuireBio™
QP7777-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010731
|
enQuireBio™
QP7777-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16020731
|
enQuireBio™
QP7777-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16030731
|
enQuireBio™
QP7777-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_009215.1 | |
Twinfilin-2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
11344 | |
Research Use Only | |
TWF2 | |
Recombinant Protein | |
E. coli | |
AHQTGIHATE ELKEFFAKAR AGSVRLIKVV IEDEQLVLGA SQEPVGRWDQ DYDRAVLPLL DAQQPCYLLY RLDSQNAQGF EWLFLAWSPD NSPVRLKMLY AATRATVKKE FGGGHIKDEL FGTVKDDLSF AGYQKHLSSC AAPAPLTSAE RELQQIRINE VKTEISVESK HQTLQGLAFP LQPEAQRALQ QLKQKMVNYI QMKLDLERET IELVHTEPTD VAQLPSRVPR DAARYHFFLY KHTHEGDPLE SVVFIYSMPG YKCSIKERML YSSCKSRLLD SVEQDFHLEI AKKIEIGDGA ELTAEFLYDE VHPKQHAFKQ AFAKPKGPGG KRGHKRLIRG PGENGDDS | |
Greater than 80% as determined by SDS-PAGE. |