missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Twinkle protein, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Twinkle protein, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9861-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001157284.1 | |
Twinkle protein, mitochondrial Protein | |
Research Use Only | |
PEO1 | |
Recombinant Protein | |
Yeast | |
APGPPRRRYR KETLQALDMP VLPVTATEIR QYLRGHGIPF QDGHSCLRAL SPFAESSQLK GQTGVTTSFS LFIDKTTGHF LCMTSLAEGS WEDFQASVEG RGDGAREGFL LSKAPEFEDS EEVRRIWNRA IPLWELPDQE EVQLADTMFG LTKVTDDTLK RFSVRYLRPA RSLVFPWFSP GGSGLRGLKL LEAKCQGDGV SYEETTIPRP SAYHNLFGLP LISRRDAEVV LTSRELDSLA LNQSTGLPTL TLPRGTTCLP PALLPYLEQF RRIVFWLGDD LRSWEAAKLF ARKLNPKRCF LVRPGDQQPR PLEALNGGFN LSRILRTALP AWHKSIVSFR QLREEVLGEL SNVEQAAGLR WSRFPDLNRI LKGHRKGELT VFTGPTGSGK TTFISEYALD LCSQGVNTLW GSFEISNVRL ARVMLTQFAE GRLEDQLDKY DHWADRFEDL PLYFMTFHGQ QSIRTVIDTM QHAVYVYDIC HVIIDNLQFM MGHEQLSTDR IAAQDYIIGV FRKFATDNNC HVTLVIHPRK EDDDKELQTA SIFGSAKASQ EADNVLILQD RKLVTGPGKR YLQVSKNRFD GDVGVFPLEF NKNSLTFSIP PKNKARLKKI KDDTGPVAKK PSSGKKGATT QNSEICSGQA PTPDQPDTSK RSK | |
Greater than 90% as determined by SDS-PAGE. |
56652 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |