missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TXNL4A Protein
A cDNA sequence encoding the TXNL4A was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 89170.00 SEK
Specifications
Name | TXNL4A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944879
|
enQuireBio™
QP13842-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924879
|
enQuireBio™
QP13842-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934879
|
enQuireBio™
QP13842-1MG |
1 mg |
89170.00 SEK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TXNL4A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TXNL4A solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 200mM NaCl and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSYMLP HLHNGWQVDQ AILSEEDRVV VIRFGHDWDP TCMKMDEVLY SIAEKVKNFA VIYLVDITEV PDFNKMYELY DPCTVMFFFR NKHIMIDLGT GNNNKINWAM EDKQEMVDII ETVYRGARKG RGLVVSPKDY STKYRY | |
Greater than 95% as determined by SDS-PAGE. |