missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human U7 snRNA-associated Sm-like protein LSm10 Protein
A DNA sequence encoding the Homo sapiens (Human) U7 snRNA-associated Sm-like protein LSm10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7867-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116270.1 | |
U7 snRNA-associated Sm-like protein LSm10 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
84967 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LSM10 | |
Recombinant Protein | |
E. coli | |
MAVSHSVKER TISENSLIIL LQGLQGRVTT VDLRDESVAH GRIDNVDAFM NIRLAKVTYT DRWGHQVKLD DLFVTGRNVR YVHIPDDVNI TSTIEQQLQI IHRVRNFGGK GQGRWEFPPK NCK |