missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2F Protein
A DNA sequence encoding the Homo sapiens (Human) UBE2F, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001265234.1 |
---|---|
Gene ID (Entrez) | 140739 |
Name | UBE2F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051831
|
enQuireBio™
QP7958-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071831
|
enQuireBio™
QP7958-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061831
|
enQuireBio™
QP7958-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081831
|
enQuireBio™
QP7958-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091831
|
enQuireBio™
QP7958-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001841
|
enQuireBio™
QP7958-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001265234.1 | |
UBE2F Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
140739 | |
Research Use Only | |
UBE2F | |
Recombinant Protein | |
E. coli | |
MLTLASKLKR DDGLKGSRTA ATASDSTRRV SVRDKLLVKE VAELEANLPC TCKVHFPDPN KLHCFQLTVT PDEGYYQGGK FQFETEVPDA YNMVPPKVKC LTKIWHPNIT ETGEICLSLL REHSIDGTGW APTRTLKDVV WGLNSLFTDL LNFDDPLNIE AAEHHLRDKE DFRNKVDDYI KRYAR | |
Greater than 80% as determined by SDS-PAGE. |