missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin carboxyl-terminal hydrolase 14 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin carboxyl-terminal hydrolase 14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6873-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001032411.1 | |
Ubiquitin carboxyl-terminal hydrolase 14 Protein | |
Research Use Only | |
USP14 | |
Recombinant Protein | |
E. coli | |
MPLYSVTVKW GKEKFEGVEL NTDEPPMVFK AQLFALTGVQ PARQKVMVKG GTLKDDDWGN IKIKNGMTLL MMGSADALPE EPSAKTVFVE DMTEEQLASA MELPCGLTNL GNTCYMNATV QCIRSVPELK DALKRYAGAL RASGEMASAQ YITAALRDLF DSMDKTSSSI PPIILLQFLH MAFPQFAEKG EQGQYLQQDA NECWIQMMRV LQQKLEAIED DSVKETDSSS ASAATPSKKK SLIDQFFGVE FETTMKCTES EEEEVTKGKE NQLQLSCFIN QEVKYLFTGL KLRLQEEITK QSPTLQRNAL YIKSSKISRL PAYLTIQMVR FFYKEKESVN AKVLKDVKFP LMLDMYELCT PELQEKMVSF RSKFKDLEDK KVNQQPNTSD KKSSPQKEVK YEPFSFADDI GSNNCGYYDL QAVLTHQGRS SSSGHYVSWV KRKQDEWIKF DDDKVSIVTP EDILRLSGGG DWHIAYVLLY GPRRVEIMEE ESEQ | |
Greater than 90% as determined by SDS-PAGE. |
9097 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |