missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin G76A Protein
A cDNA sequence encoding the Ubiquitin G76A was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 48005.00 SEK
Specifications
Gene ID (Entrez) | 7311; 7314; 6233; 7316 |
---|---|
Name | Ubiquitin G76A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | RPS27A; UBB; UBC; UBA52 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944969
|
enQuireBio™
QP13870-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934969
|
enQuireBio™
QP13870-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924969
|
enQuireBio™
QP13870-1MG |
1 mg |
48005.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7311; 7314; 6233; 7316 | |
Research Use Only | |
RPS27A; UBB; UBC; UBA52 | |
Human | |
Untagged | |
Greater than 95% as determined by SDS-PAGE. |
Ubiquitin G76A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
Diluted in PBS plus 5% glycerol. |