missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-like protein ISG15 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-like protein ISG15, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_056598.2 |
---|---|
Gene ID (Entrez) | 100038882 |
Name | Ubiquitin-like protein ISG15 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055901
|
enQuireBio™
QP8673-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095901
|
enQuireBio™
QP8673-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045901
|
enQuireBio™
QP8673-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075901
|
enQuireBio™
QP8673-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085901
|
enQuireBio™
QP8673-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065901
|
enQuireBio™
QP8673-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_056598.2 | |
Ubiquitin-like protein ISG15 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
100038882 | |
Research Use Only | |
G1P2 | |
Recombinant Protein | |
E. coli | |
AWDLKVKMLG GNDFLVSVTN SMTVSELKKQ IAQKIGVPAF QQRLAHQTAV LQDGLTLSSL GLGPSSTVML VVQNCSEPLS ILVRNERGHS NIYEVFLTQT VDTLKKKVSQ REQVHEDQFW LSFEGRPMED KELLGEYGLK PQCTVIKHLR LRGGGG | |
Greater than 90% as determined by SDS-PAGE. |