missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UCP1, mitochondrial brown fat Protein
A DNA sequence encoding the Homo sapiens (Human) UCP1, mitochondrial brown fat, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6868-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068605.1 | |
UCP1, mitochondrial brown fat Protein | |
Research Use Only | |
UCP1 | |
Recombinant Protein | |
E. coli | |
GGLTASDVHP TLGVQLFSAG IAACLADVIT FPLDTAKVRL QVQGECPTSS VIRYKGVLGT ITAVVKTEGR MKLYSGLPAG LQRQISSASL RIGLYDTVQE FLTAGKETAP SLGSKILAGL TTGGVAVFIG QPTEVVKVRL QAQSHLHGIK PRYTGTYNAY RIIATTEGLT GLWKGTTPNL MRSVIINCTE LVTYDLMKEA FVKNNILADD VPCHLVSALI AGFCATAMSS PVDVVKTRFI NSPPGQYKSV PNCAMKVFTN EGPTAFFKGL VPSFLRLGSW NVIMFVCFEQ LKRELSKSRQ TMDCAT | |
Greater than 90% as determined by SDS-PAGE. |
7350 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |