missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UCP1, mitochondrial brown fat Protein

A DNA sequence encoding the Homo sapiens (Human) UCP1, mitochondrial brown fat, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6868-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046420

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_068605.1
UCP1, mitochondrial brown fat Protein
Research Use Only
UCP1
Recombinant Protein
E. coli
GGLTASDVHP TLGVQLFSAG IAACLADVIT FPLDTAKVRL QVQGECPTSS VIRYKGVLGT ITAVVKTEGR MKLYSGLPAG LQRQISSASL RIGLYDTVQE FLTAGKETAP SLGSKILAGL TTGGVAVFIG QPTEVVKVRL QAQSHLHGIK PRYTGTYNAY RIIATTEGLT GLWKGTTPNL MRSVIINCTE LVTYDLMKEA FVKNNILADD VPCHLVSALI AGFCATAMSS PVDVVKTRFI NSPPGQYKSV PNCAMKVFTN EGPTAFFKGL VPSFLRLGSW NVIMFVCFEQ LKRELSKSRQ TMDCAT
Greater than 90% as determined by SDS-PAGE.
7350
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers