missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UDP-N-acetylhexosamine pyrophosphorylase Protein

A DNA sequence encoding the Homo sapiens (Human) UDP-N-acetylhexosamine pyrophosphorylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9752-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058661

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001311042.1
UDP-N-acetylhexosamine pyrophosphorylase Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
6675
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UAP1
Recombinant Protein
Yeast
MNINDLKLTL SKAGQEHLLR FWNELEEAQQ VELYAELQAM NFEELNFFFQ KAIEGFNQSS HQKNVDARME PVPREVLGSA TRDQDQLQAW ESEGLFQISQ NKVAVLLLAG GQGTRLGVAY PKGMYDVGLP SRKTLFQIQA ERILKLQQVA EKYYGNKCII PWYIMTSGRT MESTKEFFTK HKYFGLKKEN VIFFQQGMLP AMSFDGKIIL EEKNKVSMAP DGNGGLYRAL AAQNIVEDME QRGIWSIHVY CVDNILVKVA DPRFIGFCIQ KGADCGAKVV EKTNPTEPVG VVCRVDGVYQ VVEYSEISLA TAQKRSSDGR LLFNAGNIAN HFFTVPFLRD VVNVYEPQLQ HHVAQKKIPY VDTQGQLIKP DKPNGIKMEK FVFDIFQFAK KFVVYEVLRE DEFSPLKNAD SQNGKDNPTT ARHALMSLHH CWVLNAGGHF IDENGSRLPA IPRSATNGKS ETITADVNHN LKDANDVPIQ CEISPLISYA GEGLESYVAD KEFHAPLIID ENGVHELVKN GI
Videos
SDS
Documents