missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UFD1L, homolog Protein
A DNA sequence encoding the Homo sapiens (Human) UFD1L, homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7910-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001030324.2 | |
UFD1L, homolog Protein | |
Research Use Only | |
UFD1L | |
Recombinant Protein | |
E. coli | |
MFSFNMFDHP IPRVFQNRFS TQYRCFSVSM LAGPNDRSDV EKGGKIIMPP SALDQLSRLN ITYPMLFKLT NKNSDRMTHC GVLEFVADEG ICYLPHWMMQ NLLLEEGGLV QVESVNLQVA TYSKFQPQSP DFLDITNPKA VLENALRNFA CLTTGDVIAI NYNEKIYELR VMETKPDKAV SIIECDMNVD FDAPLGYKEP ERQVQHEEST EGEADHSGYA GELGFRAFSG SGNRLDGKKK GVEPSPSPIK PGDIKRGIPN YEFKLGKITF IRNSRPLVKK VEEDEAGGRF VAFSGEGQSL RKKGRKP | |
Greater than 80% as determined by SDS-PAGE. |
7353 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |