missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UQCRFS1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) UQCRFS1, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8480-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005994.2 | |
UQCRFS1, mitochondrial Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
7386 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
UQCRFS1 | |
Recombinant Protein | |
E. coli | |
SHTDIKVPDF SEYRRLEVLD STKSSRESSE ARKGFSYLVT GVTTVGVAYA AKNAVTQFVS SMSASADVLA LAKIEIKLSD IPEGKNMAFK WRGKPLFVRH RTQKEIEQEA AVELSQLRDP QHDLDRVKKP EWVILIGVCT HLGCVPIANA GDFGGYYCPC HGSHYDASGR IRLGPAPLNL EVPTYEFTSD DMVIVG |