missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uridine-cytidine kinase 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Uridine-cytidine kinase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6867-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001129426.1 | |
Uridine-cytidine kinase 1 Protein | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
83549 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
UCK1 | |
Recombinant Protein | |
E. coli | |
MASAGGEDCE SPAPEADRPH QRPFLIGVSG GTASGKSTVC EKIMELLGQN EVEQRQRKVV ILSQDRFYKV LTAEQKAKAL KGQYNFDHPD AFDNDLMHRT LKNIVEGKTV EVPTYDFVTH SRLPETTVVY PADVVLFEGI LVFYSQEIRD MFHLRLFVDT DSDVRLSRRD KEVCRCDHPA RSGQYGCHQP DRAAHPGHSE W |