missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Urokinase / PLAU Protein

A DNA sequence encoding the Homo sapiens (Human) Urokinase / PLAU, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8792-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056491

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001138503.1
Urokinase / PLAU Protein
Research Use Only
PLAU
Recombinant Protein
E. coli
SNELHQVPSN CDCLNGGTCV SNKYFSNIHW CNCPKKFGGQ HCEIDKSKTC YEGNGHFYRG KASTDTMGRP CLPWNSATVL QQTYHAHRSD ALQLGLGKHN YCRNPDNRRR PWCYVQVGLK PLVQECMVHD CADGKKPSSP PEELKFQCGQ KTL
Greater than 90% as determined by SDS-PAGE.
5328
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers