missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human USH1C Protein
A DNA sequence encoding the Homo sapiens (Human) USH1C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8285-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001284693.1 | |
USH1C Protein | |
200 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10083 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
USH1C | |
Recombinant Protein | |
E. coli | |
MDRKVAREFR HKVDFLIEND AEKDYLYDVL RMYHQTMDVA VLVGDLKLVI NEPSRLPLFD AIRPLIPLKH QVEYDQLTPR RSRKLKEVRL DRLHPEGLGL SVRGGLEFGC GLFISHLIKG GQADSVGLQV GDEIVRINGY SISSCTHEEV INLIRTKKTV SIKVRHIGLI PVKSSPDEPL TWQYVDQFVS ESGGVRGSLG SPGNRENKEK KVFISLVGSR GLGCSISSGP IQKPGIFISH VKPGSLSAEV GLEIGDQIVE VNGVDFSNLD HKEGRELFMT DRERLAEARQ RELQRQELLM QKRLAMESNK ILQEQQEMER QRRKEIAQKA AEENERYRKE MEQIVEEEEK FKKQWEEDWG SKEQLLLPKT ITAEVHPVPL RKPKYDQGVE PELEPADDLD GGTEEQGEQD FRKYEEGFDP YSMFTPEQIM GKDVRLLRIK KEGSLDLALE GGVDSPIGKV VVSAVYERGA AERHGGIVKG DEIMAINGKI VTDYTLAEAE AALQKAWNQG GDWIDLVVAV CPPKEYDDEL TFF |