Läs mer
enQuireBio™ Recombinant Human UV excision repair protein RAD23 homolog A Protein
Produktkod. p-7153366
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16034290
Brand: enQuireBio™ QP6578ec10ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) UV excision repair protein RAD23 homolog A, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_001257291.1
Gen-ID (Entrez) 5886
Namn UV excision repair protein RAD23 homolog A Protein
Kvantitet 10 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol RAD23A
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag GST
Sekvens MAVTITLKTL QQQTFKIRME PDETVKVLKE KIEAEKGRDA FPVAGQKLIY AGKILSDDVP IRDYRIDEKN FVVVMVTKTK AGQGTSAPPE ASPTAAPESS TSFPPAPTSG MSHPPPAARE DKSPSEESAP TTSPESVSGS VPSSGSSGRE EDAASTLVTG SEYETMLTEI MSMGYERERV VAALRASYNN PHRAVEYLLT GIPGSPEPEH GSVQESQVSE QPATEAAGEN PLEFLRDQPQ FQNMRQVIQQ NPALLPALLQ QLGQENPQLL QQISRHQEQF IQMLNEPPGE LADISDVEGE VGAIGEEAPQ MNYIQVTPQE KEAIERLKAL GFPESLVIQA YFACEKNENL AANFLLSQNF DDE
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 80% as determined by SDS-PAGE.