missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human V-type proton ATPase subunit G 1 Protein
A DNA sequence encoding the Homo sapiens (Human) V-type proton ATPase subunit G 1, was expressed in the hosts and tags indicated.
1580.00 SEK - 19050.00 SEK
Specifications
Accession Number | NP_004879.1 |
---|---|
Gene ID (Entrez) | 9550 |
Name | V-type proton ATPase subunit G 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063931
|
enQuireBio™
QP8310-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003941
|
enQuireBio™
QP8310-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053931
|
enQuireBio™
QP8310-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083931
|
enQuireBio™
QP8310-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093931
|
enQuireBio™
QP8310-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073931
|
enQuireBio™
QP8310-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004879.1 | |
V-type proton ATPase subunit G 1 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9550 | |
Research Use Only | |
ATP6G | |
Recombinant Protein | |
E. coli | |
ASQSQGIQQL LQAEKRAAEK VSEARKRKNR RLKQAKEEAQ AEIEQYRLQR EKEFKAKEAA ALGSRGSCST EVEKETQEKM TILQTYFRQN RDEVLDNLLA FVCDIRPEIH ENYRING | |
Greater than 90% as determined by SDS-PAGE. |