missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VEGF-C Protein
A DNA sequence encoding the Homo sapiens (Human) VEGF-C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8577-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005420.1 | |
VEGF-C Protein | |
Research Use Only | |
VEGFC | |
Recombinant Protein | |
E. coli | |
AHYNTEILKS IDNEWRKTQC MPREVCIDVG KEFGVATNTF FKPPCVSVYR CGGCCNSEGL QCMNTSTSYL SKTLFEITVP LSQGPKPVTI SFANHTSCRC MSKLDVYRQV HSIIRR | |
Greater than 90% as determined by SDS-PAGE. |
7424 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |