missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human VTI1B Protein

A DNA sequence encoding the Homo sapiens (Human) VTI1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8206-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053271

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_006361.1
VTI1B Protein
Research Use Only
VTI1B
Recombinant Protein
E. coli
SSEHFEKLHE IFRGLHEDLQ GVPERLLGTA GTEEKKKLIR DFDEKQQEAN ETLAEMEEEL RYAPLSFRNP MMSKLRNYRK DLAKLHREVR STPLTATPGG RGDMKYGIYA VENEHMNRLQ SQRAMLLQGT ESLNRATQSI ERSHRIATET DQIGSEIIEE LGEQRDQLER TKSRLVNTSE NLSKSRKILR SMSRKVTTNK LL
Greater than 90% as determined by SDS-PAGE.
10490
200 μg
Not determined.
Cytoplasmic Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers