missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) WD repeat-containing protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6884-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060058.1 | |
WD repeat-containing protein 1 | |
Research Use Only | |
WDR5 | |
Recombinant Protein | |
E. coli | |
ATEEKKPETE AARAQPTPSS SATQSKPTPV KPNYALKFTL AGHTKAVSSV KFSPNGEWLA SSSADKLIKI WGAYDGKFEK TISGHKLGIS DVAWSSDSNL LVSASDDKTL KIWDVSSGKC LKTLKGHSNY VFCCNFNPQS NLIVSGSFDE SVRIWDVKTG KCLKTLPAHS DPVSAVHFNR DGSLIVSSSY DGLCRIWDTA SGQCLKTLID DDNPPVSFVK FSPNGKYILA ATLDNTLKLW DYSKGKCLKT YTGHKNEKYC IFANFSVTGG KWIVSGSEDN LVYIWNLQTK EIVQKLQGHT DVVISTACHP TENIIASAAL ENDKTIKLWK SDC | |
Greater than 90% as determined by SDS-PAGE. |
11091 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |