missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Wilm's Tumor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Wilm's Tumor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6890-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000369.3 | |
Wilm's Tumor 1 Protein | |
Research Use Only | |
WT1 | |
Recombinant Protein | |
Yeast | |
MGSDVRDLNA LLPAVPSLGG GGGCALPVSG AAQWAPVLDF APPGASAYGS LGGPAPPPAP PPPPPPPPHS FIKQEPSWGG AEPHEEQCLS AFTVHFSGQF TGTAGACRYG PFGPPPPSQA SSGQARMFPN APYLPSCLES QPAIRNQGYS TVTFDGTPSY GHTPSHHAAQ FPNHSFKHED PMGQQGSLGE QQYSVPPPVY GCHTPTDSCT GSQALLLRTP YSSDNLYQMT SQLECMTWNQ MNLGATLKGV AAGSSSSVKW TEGQSNHSTG YESDNHTTPI LCGAQYRIHT HGVFRGIQDV RRVPGVAPTL VRSASETSEK RPFMCAYPGC NKRYFKLSHL QMHSRKHTGE KPYQCDFKDC ERRFSRSDQL KRHQRRHTGV KPFQCKTCQR KFSRSDHLKT HTRTHTGKTS EKPFSCRWPS CQKKFARSDE LVRHHNMHQR NMTKLQLAL | |
Greater than 90% as determined by SDS-PAGE. |
7490 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |