missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human XCL1 Protein

A DNA sequence encoding the Homo sapiens (Human) XCL1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8667-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065861

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002986.1
XCL1 Protein
Research Use Only
XCL1
Recombinant Protein
E. coli
VGSEVSDKRT CVSLTTQRLP VSRIKTYTIT EGSLRAVIFI TKRGLKVCAD PQATWVRDVV RSMDRKSNTR NNMIQTKPTG TQQSTNTAVT LTG
Greater than 90% as determined by SDS-PAGE.
6375
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers