missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Zinc finger protein 160 Protein

A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 160, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6902-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066650

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001096073.1
Zinc finger protein 160 Protein
200 μg
Not determined.
Full Length of Isoform 2
Human
His
Tris-based buffer, 50% glycerol
90338
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ZNF160
Recombinant Protein
E. coli
MALTQVRLTF RDVAIEFSQE EWKCLDPAQR ILYRDVMLEN YWNLVSLGLC HFDMNIISML EEGKEPWTVK SCVKIARKPR TPECVKGVVT DLLRRWKHWL LLLGICCPKP HGRVSSRLRL SRSLGHFFHS AFATFMGVCD KRVGSIF
Videos
SDS
Documents
Certificates
Special Offers

Special Offers