missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 160 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 160, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6902-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001096073.1 | |
Zinc finger protein 160 Protein | |
Research Use Only | |
ZNF160 | |
Recombinant Protein | |
E. coli | |
MALTQVRLTF RDVAIEFSQE EWKCLDPAQR ILYRDVMLEN YWNLVSLGLC HFDMNIISML EEGKEPWTVK SCVKIARKPR TPECVKGVVT DLLRRWKHWL LLLGICCPKP HGRVSSRLRL SRSLGHFFHS AFATFMGVCD KRVGSIF | |
Greater than 90% as determined by SDS-PAGE. |
90338 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |