missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Fort Monmouth / 1 / 1947) NucleoProtein
A DNA sequence encoding the Influenza A virus Influenza A H1N1 (strain A/Fort Monmouth/1/1947) Nucleoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7158-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H1N1 (Fort Monmouth / 1 / 1947) NucleoProtein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMVGGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELIL YDKEEIRRIW RQANNGDDAT AGLTHIMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRIAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEREGYSLVG IDPFKLLQNS HVYSLIRPNE NPAHKSQLVW MACNSAAFED LRLSSFIRGT KVVPRGRLST RGIQIASNEN MDTMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISIQP TFSVQRNLPF DKTTIMAAFT GNAEGRTSDM RAEIIRMMEN ARPEEVSFQG RGVFELSDER AANPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |