missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein
A DNA sequence encoding the Influenza A virus (strain A/Hong Kong/1/1968 H3N2) Influenza A H3N2 (A/Hong Kong/1/1968) Nucleoprotein / NP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7093-500ug
Additional Details : Weight : 0.01000kg
Specifications
Influenza A H3N2 (Hong Kong / 1 / 1968) Nucleoprotein / NP Protein | |
Research Use Only | |
NP | |
Recombinant Protein | |
E. coli | |
MASQGTKRSY EQMETDGERQ NATEIRASVG KMIDGIGRFY IQMCTELKLS DYEGRLIQNS LTIERMVLSA FDERRNKYLE EHPSAGKDPK KTGGPIYKRV DGKWMRELVL YDKEEIRRIW RQANNGDDAT AGLTHMMIWH SNLNDTTYQR TRALVRTGMD PRMCSLMQGS TLPRRSGAAG AAVKGVGTMV MELIRMIKRG INDRNFWRGE NGRKTRSAYE RMCNILKGKF QTAAQRAMMD QVRESRNPGN AEIEDLIFLA RSALILRGSV AHKSCLPACV YGPAVASGYD FEKEGYSLVG IDPFKLLQNS QVYSLIRPNE NPAHKSQLVW MACNSAAFED LRVLSFIRGT KVSPRGKLST RGVQIASNEN MDAMESSTLE LRSRYWAIRT RSGGNTNQQR ASAGQISVQP AFSVQRNLPF DKPTIMAAFT GNTEGRTSDM RAEIIRMMEG AKPEEMSFQG RGVFELSDEK AANPIVPSFD MSNEGSYFFG DNAEEYDN | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |