missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein Product Code.: 16017281

enQuireBio™ Recombinant Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein

Product Code. 16017281
100 μg, 100µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16017281

Brand: enQuireBio™ QP8948ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Influenza A virus (A/chicken/Jiangsu/1021/2013(H7N9)) Influenza A H7N9 (A/Shanghai/02/2013) Hemagglutinin, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_009118475.1
Gene ID (Entrez) 23104227
Name Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol HA
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Influenza
Species E. coli
Protein Tag His-SUMO
Sequence DKICLGHHAV SNGTKVNTLT ERGVEVVNAT ETVERTNIPR ICSKGKRTVD LGQCGLLGTI TGPPQCDQFL EFSADLIIER REGSDVCYPG KFVNEEALRQ ILRESGGIDK EAMGFTYSGI RTNGATSACR RSGSSFYAEM KWLLSNTDNA AFPQMTKSYK NTRKSPALIV WGIHHSVSTA EQTKLYGSGN KLVTVGSSNY QQSFVPSPGA RPQVNGLSGR IDFHWLMLNP NDTVTFSFNG AFIAPDRASF LRGKSMGIQS GVQVDANCEG DCYHSGGTII SNLPFQNIDS RAVGKCPRYV KQRSLLLATG MKNVPEIPKG RGLFGAIAGF IENGWEGLID GWYGFRHQNA QGEGTAADYK STQSAIDQIT GKLNRLIEKT NQQFELIDNE FNEVEKQIGN VINWTRDSIT EVWSYNAELL VAMENQHTID LADSEMDKLY ERVKRQLREN AEEDGTGCFE IFHKCDDDCM ASIRNNTYDH SKYREEAMQN RIQIDPVKLS SGYKDV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Influenza A H7N9 (Shanghai / 02 / 2013) Hemagglutinin Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.