missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza-B Ohio Protein
A cDNA sequence encoding the Influenza-B Ohio was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 30850.00 SEK
Specifications
Name | Influenza-B Ohio Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970429
|
enQuireBio™
QP12441-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960429
|
enQuireBio™
QP12441-10UG |
10 μg |
3405.00 SEK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950429
|
enQuireBio™
QP12441-100UG |
100 μg |
30850.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza-B Ohio Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant B/Ohio/01/05 solution contains 30mM Sodium phosphate, pH 7.4 and 50mM NaCl. |