Learn More
enQuireBio™ Recombinant Influenza H1N1 Solomon Islands Protein
A cDNA sequence encoding the H1N1 Solomon Islands was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12078-50ug
Informations supplémentaires : Weight : 0.01000kg
Spécification
Influenza H1N1 Solomon Islands Protein | |
Research Use Only | |
Recombinant Protein | |
Influenza H1N1 | |
The Recombinant H1N1 A/Solomon Islands/03/2006 solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |