missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H3N2 Wisconsin Protein
A cDNA sequence encoding the H3N2 Wisconsin was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 34280.00 SEK
Specifications
Name | Influenza H3N2 Wisconsin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Influenza |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969338
|
enQuireBio™
QP12091-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979338
|
enQuireBio™
QP12091-50UG |
50 μg |
2405.00 SEK
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949338
|
enQuireBio™
QP12091-100UG |
100 μg |
30850.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959338
|
enQuireBio™
QP12091-1MG |
1 mg |
34280.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Influenza H3N2 Wisconsin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Influenza H3N2 | |
The Recombinant H3N2 A/Wisconsin/67/05 solution contains 10mM Sodium phosphate, pH 7.2 and 150mM Sodium Chloride. |