Learn More
enQuireBio™ Recombinant M. tuberculosis Immunogenic protein MPT63 Protein
A DNA sequence encoding the Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh) Immunogenic protein MPT63, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8820-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_003409684.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
mpt63 | |
Recombinant Protein | |
E. coli | |
AYPITGKLGS ELTMTDTVGQ VVLGWKVSDL KSSTAVIPGY PVAGQVWEAT ATVNAIRGSV TPAVSQFNAR TADGINYRVL WQAAGPDTIS GATIPQGEQS TGKIYFDVTG PSPTIVAMNN GMEDLLIWEP |
M. tuberculosis Immunogenic protein MPT63 Protein | |
50 μg | |
Not determined. | |
Full Length | |
M. tuberculosis | |
His | |
Tris-based buffer, 50% glycerol |